Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112293.5 | NP_188052.2 | 11 - 12 | 1571 | 87 | |
NM_202582.1 | NP_974311.1 | 10 - 11 | 1627 | 87 | ||
NM_202583.1 | NP_974312.1 | 11 - 12 | 1495 | 87 | ||
GenBank mRNA | AY518292.1 | - | 11 - 12 | 1565 | 87 | |
BT046202.1 | - | 11 - 12 | 1327 | 87 | ||
FJ708721.1 | - | 11 - 12 | 1327 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |