Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203955.1 | NP_001190884.1 | 6 - 7 | 593 | 84 | |
NM_001342121.1 | NP_001328314.1 | 4 - 5 | 667 | 84 | ||
NM_119341.2 | NP_194918.2 | 4 - 5 | 667 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |