Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036691.1 | NP_001031768.1 | 9 - 10 | 1387 | 149 | |
NM_001125620.1 | NP_001119092.1 | 9 - 10 | 1387 | 149 | ||
NM_119370.5 | NP_194945.3 | 9 - 10 | 1437 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |