Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125626.1 | NP_001119098.1 | 4 - 5 | 670 | 75 | |
NM_119404.3 | NP_567895.1 | 4 - 5 | 883 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |