Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203962.1 | NP_001190891.1 | 12 - 13 | 2518 | 66 | |
NM_119407.3 | NP_567896.1 | 13 - 14 | 2460 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |