Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIFY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342158.1 | NP_001328277.1 | 3 - 4 | 834 | 71 | |
NM_119409.4 | NP_567898.1 | 3 - 4 | 843 | 71 | ||
GenBank mRNA | BT006431.1 | - | 3 - 4 | 678 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |