Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119436.3 | NP_195009.1 | 4 - 5 | 512 | 86 | |
GenBank mRNA | AJ854183.1 | - | 4 - 5 | 398 | 86 | |
AK221608.1 | - | 4 - 5 | 502 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |