Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMMA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119448.4 | NP_195020.1 | 7 - 8 | 1461 | 91 | |
GenBank mRNA | AF370160.1 | - | 7 - 8 | 1173 | 91 | |
AF424619.1 | - | 7 - 8 | 1171 | 91 | ||
AY059104.1 | - | 7 - 8 | 1099 | 91 | ||
AY133531.1 | - | 7 - 8 | 1099 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |