Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XI-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203968.1 | NP_001190897.1 | 23 - 24 | 3125 | 94 | |
NM_001203969.1 | NP_001190898.1 | 23 - 24 | 3125 | 94 | ||
NM_001342203.1 | NP_001328260.1 | 23 - 24 | 3147 | 94 | ||
NM_119474.5 | NP_195046.3 | 23 - 24 | 3159 | 94 | ||
GenBank mRNA | AK228969.1 | - | 23 - 24 | 3146 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |