Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119505.3 | NP_195077.1 | 3 - 4 | 1138 | 98 | |
GenBank mRNA | AF361798.1 | - | 3 - 4 | 1062 | 98 | |
AY124858.1 | - | 3 - 4 | 1013 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |