Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342217.1 | NP_001320125.1 | 3 - 4 | 1540 | 69 | |
NM_119505.3 | NP_195077.1 | 4 - 5 | 1380 | 69 | ||
GenBank mRNA | AF361798.1 | - | 4 - 5 | 1304 | 69 | |
AY124858.1 | - | 4 - 5 | 1255 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |