Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342263.1 | NP_001329319.1 | 6 - 7 | 1213 | 73 | |
NM_001342264.1 | NP_001329320.1 | 6 - 7 | 1068 | 73 | ||
NM_119588.6 | NP_567962.1 | 7 - 8 | 1264 | 73 | ||
NM_202950.3 | NP_974679.1 | 7 - 8 | 1072 | 73 | ||
GenBank mRNA | AY054633.1 | - | 7 - 8 | 915 | 73 | |
AY081532.1 | - | 7 - 8 | 891 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |