Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342289.1 | NP_001328947.1 | 10 - 11 | 1833 | 92 | |
NM_001342290.1 | NP_001328948.1 | 10 - 11 | 1700 | 92 | ||
NM_001342291.1 | NP_001328949.1 | 11 - 12 | 1645 | 92 | ||
NM_001342292.1 | NP_001328950.1 | 10 - 11 | 1596 | 92 | ||
NM_119623.5 | NP_195183.2 | 11 - 12 | 1742 | 92 | ||
GenBank mRNA | AK227728.1 | - | 11 - 12 | 1681 | 92 | |
BT003159.1 | - | 8 - 9 | 1441 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |