Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342297.1 | NP_001329728.1 | 8 - 9 | 927 | 117 | |
NM_119630.4 | NP_195190.1 | 8 - 9 | 1173 | 117 | ||
GenBank mRNA | AY099868.1 | - | 8 - 9 | 976 | 117 | |
BT003419.1 | - | 8 - 9 | 873 | 117 | ||
D29017.1 | - | 8 - 9 | 1087 | 117 | ||
U79159.1 | - | 8 - 9 | 1000 | 117 | ||
X86692.1 | - | 986 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |