Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198471.1 | NP_001185400.1 | 7 - 8 | 1228 | 59 | |
NM_106154.4 | NP_565097.1 | 6 - 7 | 1378 | 59 | ||
NM_179557.4 | NP_849888.1 | 6 - 7 | 1378 | 59 | ||
GenBank mRNA | AF318307.1 | - | 6 - 7 | 1063 | 59 | |
AF419598.1 | - | 6 - 7 | 1142 | 59 | ||
AY054196.1 | - | 6 - 7 | 1223 | 59 | ||
AY097344.1 | - | 6 - 7 | 976 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |