Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334865.1 | NP_001321029.1 | 13 - 14 | 1771 | 75 | |
NM_106546.3 | NP_178018.1 | 13 - 14 | 1875 | 75 | ||
NM_179572.2 | NP_849903.1 | 12 - 13 | 1713 | 75 | ||
GenBank mRNA | AF360147.1 | - | 12 - 13 | 1703 | 75 | |
AY052358.1 | - | 13 - 14 | 1866 | 75 | ||
AY142678.1 | - | 12 - 13 | 1629 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |