Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126508.5 | NP_565313.1 | 2 | 1087 | 73 | |
NM_179604.2 | NP_849935.1 | 1 | 1234 | 73 | ||
GenBank mRNA | AF083754.1 | - | 2 | 795 | 73 | |
AY094449.1 | - | 2 | 825 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |