Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179667.3 | NP_849998.1 | 1 - 2 | 527 | 66 | |
GenBank mRNA | AK226880.1 | - | 1 - 2 | 449 | 66 | |
AY091071.1 | - | 1 - 2 | 448 | 66 | ||
AY133858.1 | - | 1 - 2 | 74 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |