Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179667.3 | NP_849998.1 | 7 - 8 | 1988 | 83 | |
GenBank mRNA | AK226880.1 | - | 7 - 8 | 1910 | 83 | |
AY091071.1 | - | 7 - 8 | 1909 | 83 | ||
AY133858.1 | - | 7 - 8 | 1535 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |