Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179749.3 | NP_850080.1 | 5 - 6 | 700 | 85 | |
NM_179750.3 | NP_850081.1 | 6 - 7 | 543 | 85 | ||
GenBank mRNA | AK119091.1 | - | 5 - 6 | 469 | 85 | |
BT003734.1 | - | 5 - 6 | 429 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |