Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036358.2 | NP_001031435.1 | 5 - 6 | 1168 | 105 | |
NM_001336153.1 | NP_001323735.1 | 5 - 6 | 901 | 105 | ||
NM_001336154.1 | NP_001323736.1 | 5 - 6 | 642 | 105 | ||
NM_001336155.1 | NP_001323737.1 | 5 - 6 | 1027 | 105 | ||
NM_001336156.1 | NP_001323738.1 | 5 - 6 | 950 | 105 | ||
NM_179784.2 | NP_850115.1 | 4 - 5 | 1137 | 105 | ||
GenBank mRNA | AY065457.1 | - | 4 - 5 | 689 | 105 | |
BT004417.1 | - | 4 - 5 | 519 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |