Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124933.1 | NP_001118405.1 | 5 - 6 | 787 | 111 | |
NM_128439.3 | NP_180446.1 | 5 - 6 | 1103 | 111 | ||
NM_179794.2 | NP_850125.1 | 5 - 6 | 787 | 111 | ||
GenBank mRNA | BT002436.1 | - | 5 - 6 | 792 | 111 | |
BT006602.1 | - | 5 - 6 | 693 | 111 | ||
U89272.1 | - | 5 - 6 | 729 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |