Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179839.3 | NP_850170.1 | 21 - 22 | 3096 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |