Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112402.3 | NP_188154.1 | 5 - 6 | 1406 | 99 | |
GenBank mRNA | AF361806.1 | - | 5 - 6 | 1308 | 99 | |
AY059155.1 | - | 5 - 6 | 1189 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |