Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDALPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112443.3 | NP_188194.1 | 2 - 3 | 331 | 67 | |
GenBank mRNA | AK226887.1 | - | 2 - 3 | 171 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |