Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035631.2 | NP_001030708.1 | 17 - 18 | 1100 | 117 | |
NM_001338192.1 | NP_001326807.1 | 11 - 12 | 752 | 117 | ||
NM_112465.4 | NP_188216.2 | 19 - 20 | 1214 | 117 | ||
GenBank mRNA | AY120750.1 | - | 19 - 20 | 1213 | 117 | |
BT001207.1 | - | 19 - 20 | 1172 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |