Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035631.2 | NP_001030708.1 | 24 - 25 | 1436 | 127 | |
NM_001338191.1 | NP_001326809.1 | 18 - 19 | 1109 | 127 | ||
NM_001338192.1 | NP_001326807.1 | 18 - 19 | 1088 | 127 | ||
NM_001338193.1 | NP_001326808.1 | 16 - 17 | 975 | 127 | ||
NM_112465.4 | NP_188216.2 | 26 - 27 | 1550 | 127 | ||
GenBank mRNA | AY120750.1 | - | 26 - 27 | 1549 | 127 | |
BT001207.1 | - | 26 - 27 | 1508 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |