Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202982.2 | NP_001189911.1 | 9 - 10 | 2294 | 78 | |
NM_112505.4 | NP_566543.1 | 9 - 10 | 2555 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |