Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112594.2 | NP_188343.1 | 8 - 9 | 1323 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |