Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112631.3 | NP_566580.1 | 3 - 4 | 705 | 92 | |
NM_202599.2 | NP_974328.1 | 1 - 2 | 954 | 92 | ||
GenBank mRNA | BT029495.1 | - | 1 - 2 | 91 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |