Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112640.5 | NP_188387.1 | 3 - 4 | 529 | 78 | |
GenBank mRNA | AY054274.1 | - | 3 - 4 | 500 | 78 | |
AY669802.1 | - | 3 - 4 | 422 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |