Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC64-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338317.1 | NP_001325422.1 | 4 - 5 | 692 | 89 | |
NM_112678.4 | NP_188424.2 | 4 - 5 | 737 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |