Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112681.3 | NP_188427.2 | 10 - 11 | 1512 | 80 | |
GenBank mRNA | AF197940.1 | - | 10 - 11 | 1498 | 80 | |
AF367299.1 | - | 10 - 11 | 1437 | 80 | ||
AY058175.1 | - | 10 - 11 | 1512 | 80 | ||
AY091683.1 | - | 10 - 11 | 1297 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |