Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338319.1 | NP_001327296.1 | 5 - 6 | 1325 | 123 | |
NM_001338320.1 | NP_001327298.1 | 5 - 6 | 1325 | 123 | ||
NM_001338321.1 | NP_001327297.1 | 5 - 6 | 1245 | 123 | ||
NM_112686.4 | NP_566595.1 | 5 - 6 | 845 | 123 | ||
NM_202602.2 | NP_974331.1 | 4 - 5 | 1368 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |