Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112690.4 | NP_188436.1 | 5 - 6 | 662 | 141 | |
GenBank mRNA | AK316840.1 | - | 5 - 6 | 565 | 141 | |
AK316900.1 | - | 5 - 6 | 566 | 141 | ||
AY084864.1 | - | 5 - 6 | 564 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |