Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112690.4 | NP_188436.1 | 9 - 10 | 1284 | 88 | |
GenBank mRNA | AK316840.1 | - | 9 - 10 | 1187 | 88 | |
AK316900.1 | - | 9 - 10 | 1188 | 88 | ||
AY084864.1 | - | 9 - 10 | 1186 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |