Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112733.3 | NP_566611.1 | 3 - 4 | 628 | 107 | |
GenBank mRNA | AY056225.1 | - | 3 - 4 | 572 | 107 | |
AY142680.1 | - | 3 - 4 | 396 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |