Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUC-L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338347.1 | NP_001325664.1 | 10 - 11 | 1059 | 71 | |
NM_001338348.1 | NP_001325663.1 | 11 - 12 | 1092 | 71 | ||
NM_112747.3 | NP_188491.1 | 12 - 13 | 1124 | 71 | ||
GenBank mRNA | DQ446672.1 | - | 12 - 13 | 1115 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |