Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGSIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112752.3 | NP_566615.1 | 2 - 3 | 339 | 125 | |
GenBank mRNA | AY056133.1 | - | 2 - 3 | 339 | 125 | |
AY142690.1 | - | 2 - 3 | 315 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |