Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112791.6 | NP_188535.4 | 30 - 31 | 7666 | 137 | |
GenBank mRNA | DQ399530.1 | - | 29 - 30 | 7466 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |