Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203003.1 | NP_001189932.1 | 10 - 11 | 1983 | 73 | |
NM_112804.5 | NP_188548.2 | 10 - 11 | 2212 | 73 | ||
GenBank mRNA | AY090240.1 | - | 10 - 11 | 1988 | 73 | |
AY091051.1 | - | 10 - 11 | 1986 | 73 | ||
BT002372.1 | - | 10 - 11 | 1922 | 73 | ||
BT006362.1 | - | 10 - 11 | 1922 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |