Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203004.1 | NP_001189933.1 | 11 - 12 | 1564 | 117 | |
NM_001338387.1 | NP_001325757.1 | 11 - 12 | 1545 | 117 | ||
NM_112808.4 | NP_188552.3 | 11 - 12 | 1545 | 117 | ||
GenBank mRNA | AB250666.1 | - | 11 - 12 | 1520 | 117 | |
DQ912973.1 | - | 11 - 12 | 1458 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |