Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112861.4 | NP_188605.1 | 7 - 8 | 987 | 72 | |
GenBank mRNA | AF446892.1 | - | 7 - 8 | 834 | 72 | |
AJ271732.1 | - | 7 - 8 | 883 | 72 | ||
AK226525.1 | - | 7 - 8 | 903 | 72 | ||
AY052676.1 | - | 7 - 8 | 890 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |