Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338447.1 | NP_001326463.1 | 3 - 4 | 588 | 75 | |
NM_112893.4 | NP_188637.2 | 3 - 4 | 586 | 75 | ||
GenBank mRNA | BT029008.1 | - | 3 - 4 | 432 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |