Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DELTA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112912.3 | NP_188656.1 | 6 - 7 | 1190 | 65 | |
GenBank mRNA | AB105106.1 | - | 6 - 7 | 1091 | 65 | |
AF521661.1 | - | 6 - 7 | 1094 | 65 | ||
AY120765.1 | - | 6 - 7 | 1117 | 65 | ||
BT000949.1 | - | 6 - 7 | 1055 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |