Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342345.1 | NP_001329080.1 | 5 - 6 | 1643 | 130 | |
NM_119697.4 | NP_195257.1 | 5 - 6 | 1776 | 130 | ||
GenBank mRNA | AK227104.1 | - | 5 - 6 | 1395 | 130 | |
U31834.1 | - | 4 - 5 | 1269 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |