Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342345.1 | NP_001329080.1 | 7 - 8 | 2041 | 92 | |
NM_119697.4 | NP_195257.1 | 7 - 8 | 2174 | 92 | ||
GenBank mRNA | AK227104.1 | - | 7 - 8 | 1793 | 92 | |
U31834.1 | - | 6 - 7 | 1667 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |