Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119708.3 | NP_195268.2 | 2 - 3 | 301 | 76 | |
GenBank mRNA | BT022119.1 | - | 2 - 3 | 297 | 76 | |
BT025661.1 | - | 2 - 3 | 293 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |