Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119743.4 | NP_195302.1 | 4 - 5 | 670 | 87 | |
GenBank mRNA | AY078973.1 | - | 4 - 5 | 526 | 87 | |
AY091671.1 | - | 4 - 5 | 475 | 87 | ||
U26945.1 | - | 4 - 5 | 509 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |