Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085030.1 | NP_001078499.1 | 3 - 4 | 793 | 90 | |
NM_119752.4 | NP_195311.1 | 4 - 5 | 679 | 90 | ||
GenBank mRNA | AY065262.1 | - | 4 - 5 | 608 | 90 | |
AY085750.1 | - | 4 - 5 | 608 | 90 | ||
AY117348.1 | - | 4 - 5 | 471 | 90 | ||
U46925.1 | - | 4 - 5 | 598 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |